MilliporeSigma
  • Possible role of fibroblast growth factor 21 on atherosclerosis via amelioration of endoplasmic reticulum stress-mediated apoptosis in apoE(-/-) mice.
Possible role of fibroblast growth factor 21 on atherosclerosis via amelioration of endoplasmic reticulum stress-mediated apoptosis in apoE(-/-) mice.
Heart and vessels (2014-08-06)
Xi Wu, Yong-Fen Qi, Jin-Rui Chang, Wei-Wei Lu, Jin-Sheng Zhang, Shao-Ping Wang, Shu-Juan Cheng, Ming Zhang, Qian Fan, Yuan Lv, Hui Zhu, Man-Kun Xin, Yun Lv, Jing-Hua Liu
ABSTRACT
Fibroblast growth factor 21 (FGF-21) is an endocrine factor that can be secreted into circulation by the liver. FGF-21 takes part in metabolic actions and is thought to be a promising candidate for the treatment of diabetes. However, the role of FGF-21 in atherosclerosis is unknown. In this study, apoE(-/-) mice were fed an atherogenic diet for 4xa0weeks with and without subcutaneous injections of FGF-21. ApoE(-/-) mice fed an atherogenic diet showed hyperlipidemia, a large plaque area in aortas and increased vessel wall thickness. Plasma FGF-21 content and protein level of FGF receptor 1 (FGFR1) in aortas was greater in apoE(-/-) than C57BL/6J mice. Exogenous FGF-21 treatment significantly ameliorated dyslipidemia in apoE(-/-) mice. FGF-21-treated apoE(-/-) mice showed reduced number of aortic plaques and plaque area as well as reduced number of TUNEL-positive cells. Protein levels of the endoplasmic reticulum stress markers glucose-regulated protein 94, caspase-12 and C/EBP homologous protein were reduced by 34.5, 31.4 and 26.5xa0%, respectively, in apoE(-/-) mice. Endogenous expression of FGF-21 and its receptor FGFR1 were upregulated in apoE(-/-) mice, and exogenous administration of FGF-21 ameliorated the atherogenic-induced dyslipidemia and vascular atherosclerotic lesions. FGF-21 protecting against atherosclerosis might be in part by its inhibitory effects on endoplasmic reticulum stress-mediated apoptosis.