MilliporeSigma
  • Detection of extranodal spread in head and neck cancer with [18F]FDG PET and MRI: improved accuracy?
Detection of extranodal spread in head and neck cancer with [18F]FDG PET and MRI: improved accuracy?
The quarterly journal of nuclear medicine and molecular imaging : official publication of the Italian Association of Nuclear Medicine (AIMN) [and] the International Association of Radiopharmacology (IAR), [and] Section of the Society of... (2014-01-03)
W L Lodder, W V Vogel, C A Lange, O Hamming-Vrieze, M L Van Velthuysen, F A Pameijer, A J Balm, M W Van Den Brekel
ABSTRACT
Preoperative detection of extranodal spread (ENS) in head and neck cancer can have important consequences for patient management. The aim of this study was to determine whether 18-fluorodeoxyglucose positron emission tomography ([18F]FDG PET) or a combination with Magnetic Resonance Imaging (MRI) could more accurately predict ENS, especially with the near availability of fully integrated [18F]FDG PET/MRI scanners. In retrospective cohort design a total of twelve patients, with 18 lymphnode metastases were studied with [18F]FDG PET and MRI. Presence of ENS was scored on MRI, and [18F]FDG PET images using a SUV max cut-off point of 12. Histopathology results were used as reference standard. Sensitivity, specificity and accuracy were calculated. The sensitivity, specificity and accuracy of [18F]FDG PET for ENS reached 70%,100% and 83%, respectively. The mean SUVmax of ENS positive lymphnodes was 13.6 versus 8.7 for lymphnode metastases without ENS (P=0.03). The sensitivity, specificity and accuracy of MRI for ENS were 70%, 100% and 83%, respectively. When the [18F]FDG PET and MRI findings were combined sensitivity, specificity and accuracy were 80%, 100% and 89%, respectively. Thus, accuracy increased from 83% to 89%. When there is no ENS or doubt of ENS on MRI, [18F]FDG PET seems to have additional value since it improves sensitivity and resolves uncertainty in case of high FDG uptake. This benefit needs to be confirmed prospectively in a larger cohort.
Sigma-Aldrich
Meglumine, 99.0-100.5% dry basis, meets USP testing specifications
Sigma-Aldrich
N-Methyl-D-glucamine, 99.0-100.5% (titration)