MilliporeSigma
  • PRDM1 expression via human parvovirus B19 infection plays a role in the pathogenesis of Hashimoto thyroiditis.
PRDM1 expression via human parvovirus B19 infection plays a role in the pathogenesis of Hashimoto thyroiditis.
Human pathology (2015-10-18)
Lu Wang, Wei-Ping Zhang, Li Yao, Wei Zhang, Jin Zhu, Wei-Chen Zhang, Yue-Hua Zhang, Zhe Wang, Qing-Guo Yan, Ying Guo, Lin-Ni Fan, Yi-Xiong Liu, Gao-Sheng Huang
ABSTRACT
Ectopic lymphoid follicle infiltration is a key event in Hashimoto thyroiditis (HT). Positive regulatory domain zinc finger protein 1 (PRDM1), which is induced by antigen stimulation, can regulate all lymphocyte lineages. Several groups independently demonstrated that human parvovirus B19 (PVB19) is closely associated with HT. Hence, we determined whether PRDM1 is expressed in HT thyroid tissue and whether there is any correlation between PRDM1 expression and PVB19 in the pathogenesis of HT. We detected PRDM1 expression in HT (n = 86), normal thyroid tissue (n = 30), and nontoxic nodular goiter (n = 20) samples using immunohistochemistry. We also detected PVB19 protein in HT samples in a double-blind manner and analyzed the correlation between the 2 proteins using immunofluorescence confocal detection and coimmunoprecipitation. Furthermore, we detected changes of the expression levels of PRDM1 and PVB19 in transfected primary thyroid follicular epithelial cells using real-time quantitative polymerase chain reaction. We found that PRDM1 protein is significantly highly expressed in the injured follicular epithelial cells in HT (83/86 cases) than in normal thyroid cells (0/30 cases) or in nontoxic nodular goiter cells (0/20 cases) (P < .001). In HT, the PRDM1 expression pattern was the same as that of PVB19, whereas PRDM1 and PVB19 were coexistent in the involved epithelial cells. Statistical analysis showed a significant correlation between PRDM1 and PVB19 (P < .001). In addition, primary thyroid epithelial cells also showed PRDM1 up-regulation after PVB19 NS1 transfection. Our findings suggest a previously unrecognized role of PRDM1 and PVB19 in the pathogenesis of HT.
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Hydrogen peroxide solution, SAJ first grade, ≥30.0%
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, purum p.a., ≥35% (RT)
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Luminol, 97%
Sigma-Aldrich
Luminol, ≥97% (HPLC)