MilliporeSigma
  • Multicolor luminescence patterning by photoactivation of semiconductor nanoparticle films.
Multicolor luminescence patterning by photoactivation of semiconductor nanoparticle films.
Journal of the American Chemical Society (2003-03-06)
Ying Wang, Zhiyong Tang, Miguel A Correa-Duarte, Luis M Liz-Marzán, Nicholas A Kotov
ABSTRACT
Indiscriminate adsorption of nanoparticles (NPs) significantly complicates the preparation of mesoscale NP patterns considered as enabling technology for many devices and processes. Instead of selected chemical functionalization of the substrate surface prior to the assembly of nanocolloids, the required optical properties - in our case, high quantum yield luminescence - are imparted to the layer-by-layer assembled films by spatially selected photoactivation. The films are made by sequential adsorption of a positively charged polyelectrolyte and a negatively charged CdSe/CdS aqueous dispersion with an initial quantum yield of 0.5-2%. The photoactivation process takes place in the presence of oxygen and may be accompanied by photoetching. A 50-500-fold increase in the luminescence intensity of CdSe/CdS citrate-stabilized particles (quantum yield 25-45%) after visible light illumination provides excellent pattern contrast. Micron scale luminescence patterns were produced from NPs of various CdSe core diameters with red, yellow, and green emission. It was also demonstrated that different emission colors such as orange and green can be combined in one image by taking advantage of spatially selective photoetching. The presented optical patterning technique significantly simplifies the preparation of luminescence patterns as compared to conventional methods. The high signal-to-noise ratio associated with it is essential for optical devices, information processing, and biophotonics. The most immediate use of this approach is expected in cryptography and cell monitoring.
Sigma-Aldrich
Cadmium selenide, -325 Mesh particle size, 99.99% trace metals basis, electronic grade
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 kit
Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)
$1,360.00