MilliporeSigma
  • Magnetic Resonance Imaging in Acute Ischemic Stroke Patients with Mild Symptoms: An Opportunity to Standardize Intravenous Thrombolysis.
Magnetic Resonance Imaging in Acute Ischemic Stroke Patients with Mild Symptoms: An Opportunity to Standardize Intravenous Thrombolysis.
Journal of stroke and cerebrovascular diseases : the official journal of National Stroke Association (2015-05-23)
Tyler A Brown, Marie Luby, Jignesh Shah, Dimitrios Giannakidis, Lawrence L Latour
ABSTRACT
Patients presenting with mild stroke symptoms are excluded inconsistently from intravenous (IV) thrombolysis. We aimed to compare acute magnetic resonance imaging findings in patients with mild symptoms to those with more severe deficits to identify clinically mild patients who might benefit from IV thrombolysis. We retrospectively studied consecutive stroke patients presenting with perfusion deficit who underwent time-of-flight magnetic resonance angiography within 24 hours of time last seen normal. Two raters measured the lesion volumes on diffusion-weighted imaging (DWI) and perfusion-weighted imaging (PWI) with mismatch (MM) calculated as PWI minus DWI. Occlusion site was categorized as "proximal," "distal," or "magnetic resonance angiography-negative" by consensus review. Stroke with mild symptoms was defined as admit National Institutes of Health Stroke Scale score of 4 or less. Values were reported as n (%). Ninety-one patients were included; 56 (61.5%) with nonmild and 35 (38.5%) with mild symptoms. After stratifying for occlusion site, there were no differences in PWI and MM lesion volumes for the nonmild versus mild patients (P = .34-.98 and P = .54-1, respectively). Furthermore, there was a trend for thrombolyzed mild stroke patients (88%, n = 7 of 8) to more likely have a favorable clinical outcome (discharge modified Rankin score ≤ 2) versus untreated patients (70%, n = 16 of 23). When present, conspicuous vessel occlusions in clinically mild stroke patients are concomitant with similar perfusion deficit and MM volumes in more clinically severe stroke patients. Coupled with a trend toward better outcomes in mild stroke patients who were treated with IV tissue plasminogen activator (t-PA), this could indicate that advanced imaging may be used in standardizing the way these patients are selected for IV t-PA therapy.
Product Description
Gadolinium, rod, 100mm, diameter 3.0mm, cast, 99%
Gadolinium, rod, 100mm, diameter 12.5mm, cast, 99%
Gadolinium, rod, 100mm, diameter 6.35mm, cast, 99%
Gadolinium, rod, 48mm, diameter 3.0mm, cast, 99%
Gadolinium, rod, 50mm, diameter 6.35mm, cast, 99%
Gadolinium, wire reel, 100mm, diameter 0.5mm, hard, 99.9%
Gadolinium, rod, 50mm, diameter 12.5mm, cast, 99%
Gadolinium, wire reel, 25mm, diameter 0.5mm, hard, 99.9%
Gadolinium, wire reel, 50mm, diameter 0.5mm, hard, 99.9%
Sigma-Aldrich
Gadolinium, −40 mesh, 99% trace rare earth metals basis
Sigma-Aldrich
Gadolinium, chips