MilliporeSigma
  • Stable and efficient hole transporting materials with a dimethylfluorenylamino moiety for perovskite solar cells.
Stable and efficient hole transporting materials with a dimethylfluorenylamino moiety for perovskite solar cells.
Chemical communications (Cambridge, England) (2015-05-12)
Hyeju Choi, Jin Woo Cho, Moon-Sung Kang, Jaejung Ko
ABSTRACT
Novel star-shaped hole transporting materials (HTMs) with a bis-dimethylfluorenylamino moiety have been synthesized and evaluated for high performance perovskite solar cell applications. Maximum power conversion efficiency of 14.21% has been achieved by using the HTM with a fused TPA core and the long-term stability was also shown to be comparable with that of .
Sigma-Aldrich
Chlorobenzene, SAJ special grade, ≥99.5%
Sigma-Aldrich
Chlorobenzene, SAJ first grade, ≥99.0%
Sigma-Aldrich
Chlorobenzene, anhydrous, 99.8%