MilliporeSigma
  • Suppression of neutrophil superoxide generation by BNP is attenuated in acute heart failure: a case for 'BNP resistance'.
Suppression of neutrophil superoxide generation by BNP is attenuated in acute heart failure: a case for 'BNP resistance'.
European journal of heart failure (2015-02-17)
Saifei Liu, Doan T M Ngo, Cher-Rin Chong, Anjalee T Amarasekera, Nathan E K Procter, Giovanni Licari, Rustem F Dautov, Simon Stewart, Yuliy Y Chirkov, John D Horowitz
ABSTRACT
The release of the B-type natriuretic peptide (BNP) is increased in heart failure (HF), a condition associated with oxidative stress. BNP is known to exert anti-inflammatory effects including suppression of neutrophil superoxide (O2(-)) release. However, BNP-based restoration of homeostasis in HF is inadequate, and the equivocal clinical benefit of a recombinant BNP, nesiritide, raises the possibility of attenuated response to BNP. We therefore tested the hypothesis that BNP-induced suppression of neutrophil O2(-) generation is impaired in patients with acute HF. We have recently characterized suppression of neutrophil O2(-) generation (PMA- or fMLP-stimulated neutrophil burst) by BNP as a measure of its physiological activity. In the present study, BNP response was compared in neutrophils of healthy subjects (n = 29) and HF patients (n = 45). Effects of BNP on fMLP-induced phosphorylation of the NAD(P)H oxidase subunit p47phox were also evaluated. In acute HF patients, the suppressing effect of BNP (1 µmol/L) on O2(-) generation was attenuated relative to that in healthy subjects (P < 0.05 for both PMA and fMLP). Analogously, BNP inhibited p47phox phosphorylation in healthy subjects but not in HF patients (P < 0.05). However, O2(-)-suppressing effects of the cell-permeable cGMP analogue (8-pCPT-cGMP) were preserved in acute HF. Conventional HF treatment for 5 weeks partially restored neutrophil BNP responsiveness (n = 25, P < 0.05), despite no significant decrease in plasma NT-proBNP levels. BNP inhibits neutrophil O2(-) generation by suppressing NAD(P)H oxidase assembly. This effect is impaired in acute HF patients, with partial recovery during treatment.
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC)
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
DL-Serine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98% (HPLC)
Sigma-Aldrich
DL-Serine, ≥98% (TLC)
Sigma-Aldrich
8-(4-Chlorophenylthio)-guanosine 3′,5′-cyclic monophosphate sodium salt, ≥95% (HPLC), powder