MilliporeSigma
  • A bipartite interaction between Hsp70 and CHIP regulates ubiquitination of chaperoned client proteins.
A bipartite interaction between Hsp70 and CHIP regulates ubiquitination of chaperoned client proteins.
Structure (London, England : 1993) (2015-02-17)
Huaqun Zhang, Joseph Amick, Ritu Chakravarti, Stephanie Santarriaga, Simon Schlanger, Cameron McGlone, Michelle Dare, Jay C Nix, K Matthew Scaglione, Dennis J Stuehr, Saurav Misra, Richard C Page
ABSTRACT
The ubiquitin ligase CHIP plays an important role in cytosolic protein quality control by ubiquitinating proteins chaperoned by Hsp70/Hsc70 and Hsp90, thereby targeting such substrate proteins for degradation. We present a 2.91 Å resolution structure of the tetratricopeptide repeat (TPR) domain of CHIP in complex with the α-helical lid subdomain and unstructured tail of Hsc70. Surprisingly, the CHIP-TPR interacts with determinants within both the Hsc70-lid subdomain and the C-terminal PTIEEVD motif of the tail, exhibiting an atypical mode of interaction between chaperones and TPR domains. We demonstrate that the interaction between CHIP and the Hsc70-lid subdomain is required for proper ubiquitination of Hsp70/Hsc70 or Hsp70/Hsc70-bound substrate proteins. Posttranslational modifications of the Hsc70 lid and tail disrupt key contacts with the CHIP-TPR and may regulate CHIP-mediated ubiquitination. Our study shows how CHIP docks onto Hsp70/Hsc70 and defines a bipartite mode of interaction between TPR domains and their binding partners.
Sigma-Aldrich
Heat Shock Protein 70 human, recombinant, expressed in E. coli, buffered aqueous solution, ≥90% (SDS-PAGE)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 L
Estimated to ship on March 21, 2025
$279.00