MilliporeSigma
  • Identification of a novel recurrent gain on 20q13 in chronic lymphocytic leukemia by array CGH and gene expression profiling.
Identification of a novel recurrent gain on 20q13 in chronic lymphocytic leukemia by array CGH and gene expression profiling.
Annals of oncology : official journal of the European Society for Medical Oncology (2012-01-10)
A E Rodríguez, C Robledo, J L García, M González, N C Gutiérrez, J A Hernández, V Sandoval, A García de Coca, I Recio, A Risueño, G Martín-Núñez, E García, R Fisac, J Conde, J de las Rivas, J M Hernández
ABSTRACT
The presence of genetic changes is a hallmark of chronic lymphocytic leukemia (CLL). The most common cytogenetic abnormalities with independent prognostic significance in CLL are 13q14, ATM and TP53 deletions and trisomy 12. However, CLL displays a great genetic and biological heterogeneity. The aim of this study was to analyze the genomic imbalances in CLL cytogenetic subsets from both genomic and gene expression perspectives to identify new recurrent alterations. The genomic imbalances and expression levels of 67 patients were analyzed. The novel recurrent abnormalities detected with bacterial artificial chromosome array were confirmed by FISH and oligonucleotide microarrays. In all cases, gene expression profiling was assessed. Copy number alterations were identified in 75% of cases. Overall, the results confirmed FISH studies for the regions frequently involved in CLL and also defined a new recurrent gain on chromosome 20q13.12, in 19% (13/67) of the CLL patients. Oligonucleotide expression correlated with the regions of loss or gain of genomic material, suggesting that the changes in gene expression are related to alterations in copy number. Our study demonstrates the presence of a recurrent gain in 20q13.12 associated with overexpression of the genes located in this region, in CLL cytogenetic subgroups.