MilliporeSigma
  • cDNA cloning and characterization of a novel gene encoding the MLF1-interacting protein MLF1IP.
cDNA cloning and characterization of a novel gene encoding the MLF1-interacting protein MLF1IP.
Oncogene (2004-04-30)
Silva H Hanissian, Umar Akbar, Bin Teng, Zorica Janjetovic, Anne Hoffmann, Johann K Hitzler, Norman Iscove, Kristin Hamre, Xiaoping Du, Yiai Tong, Suraj Mukatira, Jon H Robertson, Stephan W Morris
ABSTRACT
Myelodysplasia/acute myeloid leukemia (MDS/AML) is characterized by a t(3;5)(q25.1;q34) chromosomal translocation that forms a fusion gene between nucleophosmin (NPM) and MDS/myeloid leukemia factor 1 (MLF1). We identified a novel protein, MLF1-interacting protein (MLF1IP), that specifically associates with MLF1 by yeast two-hybrid analysis and in pulldown assays, and colocalizes with it in both the nuclei and cytoplasm of cells. The MLF1IP gene locus is at chromosome 4q35.1 and is composed of 14 exons spanning 75.8 kb of genomic DNA. The MLF1IP cDNA encodes a 46-kDa protein that contains two bipartite and two classical nuclear localization signals, two nuclear receptor-binding motifs (LXXLL), two leucine zippers, two PEST residues and several potential phosphorylation sites. MLF1IP transcripts are expressed in a variety of tissues (e.g. fetal liver, bone marrow, thymus and testis). MLF1IP appears to be a lineage-specific gene whose expression is confined exclusively to the CFU-E erythroid precursor cells, but not in mature erythrocytes. These observations, together with previous data demonstrating a role for MLF1 in suppressing red cell maturation, suggest a possible role for MLF1IP and MLF1 deregulation in the genesis of erythroleukemias.