MilliporeSigma
  • Untargeted Screening and Distribution of Organo-Bromine Compounds in Sediments of Lake Michigan.
Untargeted Screening and Distribution of Organo-Bromine Compounds in Sediments of Lake Michigan.
Environmental science & technology (2015-12-01)
Hui Peng, Chunli Chen, Jenna Cantin, David M V Saunders, Jianxian Sun, Song Tang, Garry Codling, Markus Hecker, Steve Wiseman, Paul D Jones, An Li, Karl J Rockne, Neil C Sturchio, John P Giesy
ABSTRACT
Previously unreported natural and synthetic organo-bromine compounds (NSOBCs) have been found to contribute more than 99% of total organic bromine (TOB) in environmental matrices. We recently developed a novel untargeted method (data-independent precursor isolation and characteristic fragment, DIPIC-Frag) and identified ∼2000 NSOBCs in two sediments from Lake Michigan. In this study, this method was used to investigate the distributions of these NSOBCs in 23 surficial samples and 24 segments of a sediment core from Lake Michigan. NSOBCs were detected in all 23 surficial samples and exhibited 10- to 100-fold variations in peak abundance among locations. The pattern of distributions of NSOBCs was correlated with depth of the water column (r(2) = 0.61, p < 0.001). Hierarchical cluster analysis showed that sediments in close proximity exhibited similar profiles of NSOBCs. Distributions of NSOBCs in 24 segments of a sediment core dated from 1766 to 2008 were investigated, and samples from similar depths exhibited similar profiles of NSOBCs. NSOBCs were grouped into four clusters (soft-cluster analysis) with different temporal trends of abundances. 515 and 768 of the NSOBCs were grouped into cluster 1 and cluster 3 with increasing temporal trends, especially since 1950, indicating that abundances of these compounds might have been affected by human activities.
Sigma-Aldrich
5-Bromoindole, 99%