Merck
  • Unprecedented Spectroscopic and Computational Evidence for Allenyl and Propargyl Titanocene(IV) Complexes: Electrophilic Quenching of Their Metallotropic Equilibrium.
Unprecedented Spectroscopic and Computational Evidence for Allenyl and Propargyl Titanocene(IV) Complexes: Electrophilic Quenching of Their Metallotropic Equilibrium.
Chemistry (Weinheim an der Bergstrasse, Germany) (2016-01-21)
Ana Belén Ruiz-Muelle, Pascual Oña-Burgos, Manuel A Ortuño, J Enrique Oltra, Ignacio Rodríguez-García, Ignacio Fernández
ABSTRACT
The synthesis and structural characterization of allenyl titanocene(IV) [TiClCp2 (CH=C=CH2 )] 3 and propargyl titanocene(IV) [TiClCp2 (CH2 -C≡C-(CH2 )4 CH3 )] 9 have been described for the first time. Advanced NMR methods including diffusion NMR methods (diffusion pulsed field gradient stimulated spin echo (PFG-STE) and DOSY) have been applied and established that these organometallics are monomers in THF solution with hydrodynamic radii (from the Stokes-Einstein equation) of 3.5 and 4.1 Å for 3 and 9, respectively. Full (1) H, (13) C, Δ(1) H, and Δ(13) C NMR data are given, and through the analysis of the Ramsey equation, the first electronic insights into these derivatives are provided. In solution, they are involved in their respective metallotropic allenyl-propargyl equilibria that, after quenching experiments with aromatic and aliphatic aldehydes, ketones, and protonating agents, always give the propargyl products P (when carbonyls are employed), or allenyl products A (when a proton source is added) as the major isomers. In all the cases assayed, the ratio of products suggests that the metallotropic equilibrium should be faster than the reactions of 3 and 9 with electrophiles. Indeed, DFT calculations predict lower Gibbs energy barriers for the metallotropic equilibrium, thus confirming dynamic kinetic resolution.
Sigma-Aldrich
3-Fluorobenzaldehyde, 97%