MilliporeSigma
Fragrance material review on geranyl linalool.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2008-07-22)
A Lapczynski, S P Bhatia, C S Letizia, A M Api
ABSTRACT
A toxicologic and dermatologic review of geranyl linalool when used as a fragrance ingredient is presented.
Sigma-Aldrich
Geranyllinalool, technical, ≥90% (GC)