MilliporeSigma
  • Porcine aminopeptidase N binds to F4+ enterotoxigenic Escherichia coli fimbriae.
Porcine aminopeptidase N binds to F4+ enterotoxigenic Escherichia coli fimbriae.
Veterinary research (2016-02-10)
Pengpeng Xia, Yiting Wang, Congrui Zhu, Yajie Zou, Ying Yang, Wei Liu, Philip R Hardwidge, Guoqiang Zhu
ABSTRACT
F4(+) enterotoxigenic Escherichia coli (ETEC) strains cause diarrheal disease in neonatal and post-weaned piglets. Several different host receptors for F4 fimbriae have been described, with porcine aminopeptidase N (APN) reported most recently. The FaeG subunit is essential for the binding of the three F4 variants to host cells. Here we show in both yeast two-hybrid and pulldown assays that APN binds directly to FaeG, the major subunit of F4 fimbriae, from three serotypes of F4(+) ETEC. Modulating APN gene expression in IPEC-J2 cells affected ETEC adherence. Antibodies raised against APN or F4 fimbriae both reduced ETEC adherence. Thus, APN mediates the attachment of F4(+) E. coli to intestinal epithelial cells.
Sigma-Aldrich
Jak3 Active human, recombinant, expressed in baculovirus infected insect cells, ≥60% (SDS-PAGE)