MilliporeSigma
  • Therapeutic silencing of HPV 16 E7 by systemic administration of siRNA-neutral DOPC nanoliposome in a murine cervical cancer model with obesity.
Therapeutic silencing of HPV 16 E7 by systemic administration of siRNA-neutral DOPC nanoliposome in a murine cervical cancer model with obesity.
Journal of B.U.ON. : official journal of the Balkan Union of Oncology (2016-02-09)
Hector Chapoy-Villanueva, Ivonne Martinez-Carlin, G Lopez-Berestein, Arturo Chavez-Reyes
ABSTRACT
To evaluate the effectiveness of a neutral DOPC nanoliposome system for the delivery of siRNA to tumor cells in an obese murine cervical cancer model. In vitro silencing of E6-E7 mRNA and E7 protein using siRNAE6 or siRNAE7 was analyzed in TC-1 cells by RT-PCR and Western blot. Silencing and antitumor capacities of siRNAE7-DOPC-nanoparticles (NP) were tested in vivo in both normal and obese mice using qPCR. These NPs were administered twice a week for 15 days and tumor volume and weight were recorded. Levels of in vitro E6-E7 silencing were 90% for mRNA and 60% for protein when siRNAE7 was used. On the other hand when siRNAE6 was used, the levels of silencing were 50% for E6-E7 mRNA and only 20% for protein. In vivo E7 mRNA silencing by siRNAE7-DOPC-NP was similar (60%) in both non-obese and obese mouse models. The therapeutic study showed a 65% decrease in tumor volume and a 57% reduction in tumor weight as compared to the control groups. There was no negative impact of obesity on the antitumor activity of siRNA-DOPC-NP in obese mice.