Merck
  • Neurochemical and behavioral effects of ciproxifan, a potent histamine H3-receptor antagonist.
Neurochemical and behavioral effects of ciproxifan, a potent histamine H3-receptor antagonist.
The Journal of pharmacology and experimental therapeutics (1998-11-10)
X Ligneau, J Lin, G Vanni-Mercier, M Jouvet, J L Muir, C R Ganellin, H Stark, S Elz, W Schunack, J Schwartz
ABSTRACT
Ciproxifan, i.e., cyclopropyl-(4-(3-1H-imidazol-4-yl)propyloxy) phenyl) ketone, belongs to a novel chemical series of histamine H3-receptor antagonists. In vitro, it behaved as a competitive antagonist at the H3 autoreceptor controlling [3H]histamine release from synaptosomes and displayed similar Ki values (0.5-1.9 nM) at the H3 receptor controlling the electrically-induced contraction of guinea pig ileum or at the brain H3 receptor labeled with [125I]iodoproxyfan. Ciproxifan displayed at least 3-orders of magnitude lower potency at various aminergic receptors studied in functional or binding tests. In vivo, measurement of drug plasma levels, using a novel radioreceptor assay in mice receiving ciproxifan p.o. or i.v., led to an oral bioavailability ratio of 62%. Oral administration of ciproxifan to mice enhanced by approximately 100% histamine turnover rate and steady state level of tele-methylhistamine with an ED50 of 0.14 mg/kg. Ciproxifan reversed the H3-receptor agonist induced enhancement of water consumption in rats with and ID50 of 0.09 +/- 0.04 mg/kg, i.p. In cats, ciproxifan (0.15-2 mg/kg, p.o.) induced marked signs of neocortical electroencephalogram activation manifested by enhanced fast-rhythms density and an almost total waking state. In rats, ciproxifan enhanced attention as evaluated in the five-choice task performed using a short stimulus duration. Ciproxifan appears to be an orally bioavailable, extremely potent and selective H3-receptor antagonist whose vigilance- and attention-promoting effects are promising for therapeutic applications in aging disorders.
Sigma-Aldrich
Ciproxifan maleate, ≥98% (HPLC), powder