MilliporeSigma
MAP kinase signalling cascade in Arabidopsis innate immunity.
Nature (2002-03-05)
Tsuneaki Asai, Guillaume Tena, Joulia Plotnikova, Matthew R Willmann, Wan-Ling Chiu, Lourdes Gomez-Gomez, Thomas Boller, Frederick M Ausubel, Jen Sheen
ABSTRACT
There is remarkable conservation in the recognition of pathogen-associated molecular patterns (PAMPs) by innate immune responses of plants, insects and mammals. We developed an Arabidopsis thaliana leaf cell system based on the induction of early-defence gene transcription by flagellin, a highly conserved component of bacterial flagella that functions as a PAMP in plants and mammals. Here we identify a complete plant MAP kinase cascade (MEKK1, MKK4/MKK5 and MPK3/MPK6) and WRKY22/WRKY29 transcription factors that function downstream of the flagellin receptor FLS2, a leucine-rich-repeat (LRR) receptor kinase. Activation of this MAPK cascade confers resistance to both bacterial and fungal pathogens, suggesting that signalling events initiated by diverse pathogens converge into a conserved MAPK cascade.