MilliporeSigma
  • Nutrient balancing of the adult worker bumblebee (Bombus terrestris) depends on the dietary source of essential amino acids.
Nutrient balancing of the adult worker bumblebee (Bombus terrestris) depends on the dietary source of essential amino acids.
The Journal of experimental biology (2015-01-27)
Daniel Stabler, Pier P Paoli, Susan W Nicolson, Geraldine A Wright
ABSTRACT
Animals carefully regulate the amount of protein that they consume. The quantity of individual essential amino acids (EAAs) obtained from dietary protein depends on the protein source, but how the proportion of EAAs in the diet affects nutrient balancing has rarely been studied. Recent research using the Geometric Framework for Nutrition has revealed that forager honeybees who receive much of their dietary EAAs from floral nectar and not from solid protein have relatively low requirements for dietary EAAs. Here, we examined the nutritional requirements for protein and carbohydrates of foragers of the buff-tailed bumblebee Bombus terrestris. By using protein (sodium caseinate) or an equimolar mixture of the 10 EAAs, we found that the intake target (nutritional optimum) of adult workers depended on the source and proportion of dietary EAAs. When bees consumed caseinate-containing diets in a range of ratios between 1:250 and 1:25 (protein to carbohydrate), they achieved an intake target (IT) of 1:149 (w/w). In contrast to those fed protein, bees fed the EAA diets had an IT more biased towards carbohydrates (1:560 w/w) but also had a greater risk of death than those fed caseinate. We also tested how the dietary source of EAAs affected free AAs in bee haemolymph. Bees fed diets near their IT had similar haemolymph AA profiles, whereas bees fed diets high in caseinate had elevated levels of leucine, threonine, valine and alanine in the haemolymph. We found that like honeybees, bumblebee workers prioritize carbohydrate intake and have a relatively low requirement for protein. The dietary source of EAAs influenced both the ratio of protein/EAA to carbohydrate and the overall amount of carbohydrate eaten. Our data support the idea that EAAs and carbohydrates in haemolymph are important determinants of nutritional state in insects.
Sigma-Aldrich
Casein sodium salt from bovine milk
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
Estimated to ship on March 22, 2025
$89.20