MilliporeSigma
Probiotic features of two oral Lactobacillus isolates.
Brazilian journal of microbiology : [publication of the Brazilian Society for Microbiology] (2012-01-01)
Gordana Zavisic, Sasa Petricevic, Zeljka Radulovic, Jelena Begovic, Natasa Golic, Ljubisa Topisirovic, Ivana Strahinic
ABSTRACT
In this study, we checked lactobacilli strains of human origin for their potential as probiotic. Samples were collected from oral mucosa of 16 healthy individuals, out of which twenty isolates were obtained and two of them were selected and identified as Lactobacillus plantarum (G1) and L. casei (G3). Both isolates exhibited antagonistic action towards pathogenic microorganisms such as Staphylococcus aureus, Pseudomonas aeruginosa, Escherichia coli, Salmonella abony, and Clostridium sporogenes, but not on the growth of Candida albicans. The bacteriocin activity against Staphylococcus aureus ATCC 6358-P was shown only by L. plantarum G1. Moreover, the isolates G1 and G3 showed good viability in the acid gastric environment and in the gut environment containing bovine bile salts. The viability of G1 and G3 isolates in the gastrointestinal tract, and the adhesion to the intestinal mucosa were also confirmed in vivo. The biochemical tests of blood samples revealed lower levels of serum triglycerides and cholesterol, as well as reduced activity of alkaline phosphatase in all lactobacilli-treated Wistar rats, compared to control ones. No toxicity for NMRI Ham mice was observed. According to our experimental results, these findings imply that L. plantarum G1 and L. casei G3 could be characterized as potential probiotics.
Product Description
Supelco
Organic Acids Kit, (individually packaged), analytical standard