Merck
  • Combined inhibition of p97 and the proteasome causes lethal disruption of the secretory apparatus in multiple myeloma cells.
Combined inhibition of p97 and the proteasome causes lethal disruption of the secretory apparatus in multiple myeloma cells.
PloS one (2013-09-27)
Holger W Auner, Anne Marie Moody, Theresa H Ward, Marianne Kraus, Enrico Milan, Philippa May, Aristeidis Chaidos, Christoph Driessen, Simone Cenci, Francesco Dazzi, Amin Rahemtulla, Jane F Apperley, Anastasios Karadimitris, Niall Dillon
ABSTRACT
Inhibition of the proteasome is a widely used strategy for treating multiple myeloma that takes advantage of the heavy secretory load that multiple myeloma cells (MMCs) have to deal with. Resistance of MMCs to proteasome inhibition has been linked to incomplete disruption of proteasomal endoplasmic-reticulum (ER)-associated degradation (ERAD) and activation of non-proteasomal protein degradation pathways. The ATPase p97 (VCP/Cdc48) has key roles in mediating both ERAD and non-proteasomal protein degradation and can be targeted pharmacologically by small molecule inhibition. In this study, we compared the effects of p97 inhibition with Eeyarestatin 1 and DBeQ on the secretory apparatus of MMCs with the effects induced by the proteasome inhibitor bortezomib, and the effects caused by combined inhibition of p97 and the proteasome. We found that p97 inhibition elicits cellular responses that are different from those induced by proteasome inhibition, and that the responses differ considerably between MMC lines. Moreover, we found that dual inhibition of both p97 and the proteasome terminally disrupts ER configuration and intracellular protein metabolism in MMCs. Dual inhibition of p97 and the proteasome induced high levels of apoptosis in all of the MMC lines that we analysed, including bortezomib-adapted AMO-1 cells, and was also effective in killing primary MMCs. Only minor toxicity was observed in untransformed and non-secretory cells. Our observations highlight non-redundant roles of p97 and the proteasome in maintaining secretory homeostasis in MMCs and provide a preclinical conceptual framework for dual targeting of p97 and the proteasome as a potential new therapeutic strategy in multiple myeloma.
Sigma-Aldrich
DBeQ, ≥98% (HPLC)