- Titania nanospheres from supercritical fluids.
Titania nanospheres from supercritical fluids.
IEE proceedings. Nanobiotechnology (2006-01-31)
J A Darr, S Kellici, I U Rehman
PMID16441166
ABSTRACT
Surfactant-coated amorphous titania nanospheres have been synthesised using templating 'water-in-supercritical carbon dioxide' emulsion droplets; the process represents a clean and controlled method for the manufacture of high-purity nanoparticles.