MilliporeSigma
  • An outwardly rectifying anionic background current in atrial myocytes from the human heart.
An outwardly rectifying anionic background current in atrial myocytes from the human heart.
Biochemical and biophysical research communications (2007-06-15)
H Li, H Zhang, J C Hancox, R Z Kozlowski
ABSTRACT
This report describes a hitherto unreported anionic background current from human atrial cardiomyocytes. Under whole-cell patch-clamp with anion-selective conditions, an outwardly rectifying anion current (I(ANION)) was observed, which was larger with iodide than nitrate, and with nitrate than chloride as charge carrier. In contrast with a previously identified background anionic current from small mammal cardiomyocytes, I(ANION) was not augmented by the pyrethroid tefluthrin (10 microM); neither was it inhibited by hyperosmolar external solution nor by DIDS (200 microM); thus I(ANION) was not due to basal activity of volume-sensitive anion channels. I(ANION) was partially inhibited by the Cl(-) channel blockers NPPB (50 microM) and Gly H-101 (30 microM). Incorporation of I(ANION) into a human atrial action potential (AP) simulation led to depression of the AP plateau, accompanied by alterations to plateau inward calcium current, and to AP shortening at 50% but not 90% of complete repolarization, demonstrating that I(ANION) can influence the human atrial AP profile.
Product Description
Supelco
Tefluthrin, PESTANAL®, analytical standard
Sigma-Aldrich
5-Nitro-2-(3-phenylpropylamino)benzoic acid, ≥98%