MilliporeSigma
Gas-chromatographic separation of stereoisomers of dipeptides.
Chirality (2006-05-10)
Ralf Pätzold, Christoph Theis, Hans Brückner
ABSTRACT
Synthetic dipeptides comprising mixtures of enantiomers, diastereomers, or sequential isomers were converted into their N-perfluoroacetyl dipeptide esters (perfluoroacetyl: trifluoroacetyl, pentafluoroacetyl, heptafluorobutyryl; ester: methyl, 1-propyl, 2-propyl, 2,2,2-trifluoroethyl) and analyzed by GC-MS on the chiral stationary phases Chirasil-L-Val and Lipodex-E using helium as carrier gas. Further, dipeptides were converted into their N-trifluoroacetyl dipeptide S-(+)-2-butyl esters and separated on achiral phenylmethyl polysiloxane column (HP-5 MS). Derivatization of dipeptides was performed at ambient temperature in order to avoid formation of the corresponding diketopiperazines. The best separation of stereoisomers was achieved with TFA and PFP methyl esters on Chirasil-L-Val.
Product Description
Supelco
Heptafluorobutyric anhydride, for GC derivatization, LiChropur, ≥99.0%
Supelco
Pentafluoropropionic anhydride, for GC derivatization, LiChropur, 99%