MilliporeSigma
  • Diagnostic and prognostic significance of glypican 5 and glypican 6 gene expression levels in gastric adenocarcinoma.
Diagnostic and prognostic significance of glypican 5 and glypican 6 gene expression levels in gastric adenocarcinoma.
Molecular and clinical oncology (2015-07-03)
Melike Dinccelik-Aslan, Guvem Gumus-Akay, Atilla Halil Elhan, Ekrem Unal, Ajlan Tukun
ABSTRACT
Gastric Cancer is one of the most common malignancies worldwide and the second most common cause of cancer-related mortality. Previous studies revealed several genetic alterations specific to gastric cancer. In this study, we aimed to investigate the diagnostic and prognostic significance of the expression levels of the glypican 5 and glypican 6 genes (