Merck
  • Discovery of Novel Anti-prion Compounds Using In Silico and In Vitro Approaches.
Discovery of Novel Anti-prion Compounds Using In Silico and In Vitro Approaches.
Scientific reports (2015-10-10)
Jae Wook Hyeon, Jiwon Choi, Su Yeon Kim, Rajiv Gandhi Govindaraj, Kyu Jam Hwang, Yeong Seon Lee, Seong Soo A An, Myung Koo Lee, Jong Young Joung, Kyoung Tai No, Jeongmin Lee
ABSTRACT
Prion diseases are associated with the conformational conversion of the physiological form of cellular prion protein (PrP(C)) to the pathogenic form, PrP(Sc). Compounds that inhibit this process by blocking conversion to the PrP(Sc) could provide useful anti-prion therapies. However, no suitable drugs have been identified to date. To identify novel anti-prion compounds, we developed a combined structure- and ligand-based virtual screening system in silico. Virtual screening of a 700,000-compound database, followed by cluster analysis, identified 37 compounds with strong interactions with essential hotspot PrP residues identified in a previous study of PrP(C) interaction with a known anti-prion compound (GN8). These compounds were tested in vitro using a multimer detection system, cell-based assays, and surface plasmon resonance. Some compounds effectively reduced PrP(Sc) levels and one of these compounds also showed a high binding affinity for PrP(C). These results provide a promising starting point for the development of anti-prion compounds.
Product Description
Millipore
Proteinase K, Lyophilized, Highly active serine protease that exhibits broad cleavage specificity on native and denatured proteins and is widely used in the purification of DNA and RNA.
SKU
Pack Size
Availability
Price
Quantity
Roche
Pefabloc® SC, powder, solubility: 100 mg/mL in aqueous buffer, suitable for blocking, suitable for protein purification