- CXCL14-Like Immunoreactivity Exists in Somatostatin-Containing Cells of Mouse Pancreas.
CXCL14-Like Immunoreactivity Exists in Somatostatin-Containing Cells of Mouse Pancreas.
Acta histochemica et cytochemica (2016-02-09)
Hirohumi Suzuki, Toshiharu Yamamoto
PMID26855449
ABSTRACT
Immunohistochemical techniques were employed to investigate the distribution of the chemokine CXCL14, in the mouse pancreas. CXCL14-immunoreactive cells were detected in the peripheral region of the pancreatic islets and were immunoreactive for somatostatin, but not for glucagon, insulin, and pancreatic polypeptide. Immunoelectron microscopy indicated that the CXCL14-like peptide and somatostatin co-existed in the secretory granules. CXCL14, secreted from somatostatin-containing cells, may modulate insulin secretion in a paracrine fashion, and play a novel role in glucose homeostasis in addition to its well-known chemotactic activities.
Sigma-Aldrich
Donkey Anti-Goat IgG Antibody, Rhodamine conjugate, Species Adsorbed, Chemicon®, from donkey
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-Pancreatic polypeptide/PPY antibody produced in goat, affinity isolated antibody, buffered aqueous solution