MilliporeSigma
  • Dopaminergic Receptors on CD4+ T Naive and Memory Lymphocytes Correlate with Motor Impairment in Patients with Parkinson's Disease.
Dopaminergic Receptors on CD4+ T Naive and Memory Lymphocytes Correlate with Motor Impairment in Patients with Parkinson's Disease.
Scientific reports (2016-09-23)
Natasa Kustrimovic, Emanuela Rasini, Massimiliano Legnaro, Raffaella Bombelli, Iva Aleksic, Fabio Blandini, Cristoforo Comi, Marco Mauri, Brigida Minafra, Giulio Riboldazzi, Vanesa Sanchez-Guajardo, Franca Marino, Marco Cosentino
ABSTRACT
Parkinson's disease (PD) is characterized by loss of dopaminergic neurons in substantia nigra pars compacta, α-synuclein (α-syn)-rich intraneuronal inclusions (Lewy bodies), and microglial activation. Emerging evidence suggests that CD4+ T lymphocytes contribute to neuroinflammation in PD. Since the mainstay of PD treatment is dopaminergic substitution therapy and dopamine is an established transmitter connecting nervous and immune systems, we examined CD4+ T naive and memory lymphocytes in PD patients and in healthy subjects (HS), with specific regard to dopaminergic receptor (DR) expression. In addition, the in vitro effects of α-syn were assessed on CD4+ T naive and memory cells. Results showed extensive association between DR expression in T lymphocytes and motor dysfunction, as assessed by UPDRS Part III score. In total and CD4+ T naive cells expression of D