Merck
  • sFRP2 in the aged microenvironment drives melanoma metastasis and therapy resistance.
sFRP2 in the aged microenvironment drives melanoma metastasis and therapy resistance.
Nature (2016-04-05)
Amanpreet Kaur, Marie R Webster, Katie Marchbank, Reeti Behera, Abibatou Ndoye, Curtis H Kugel, Vanessa M Dang, Jessica Appleton, Michael P O'Connell, Phil Cheng, Alexander A Valiga, Rachel Morissette, Nazli B McDonnell, Luigi Ferrucci, Andrew V Kossenkov, Katrina Meeth, Hsin-Yao Tang, Xiangfan Yin, William H Wood, Elin Lehrmann, Kevin G Becker, Keith T Flaherty, Dennie T Frederick, Jennifer A Wargo, Zachary A Cooper, Michael T Tetzlaff, Courtney Hudgens, Katherine M Aird, Rugang Zhang, Xiaowei Xu, Qin Liu, Edmund Bartlett, Giorgos Karakousis, Zeynep Eroglu, Roger S Lo, Matthew Chan, Alexander M Menzies, Georgina V Long, Douglas B Johnson, Jeffrey Sosman, Bastian Schilling, Dirk Schadendorf, David W Speicher, Marcus Bosenberg, Antoni Ribas, Ashani T Weeraratna
ABSTRACT
Cancer is a disease of ageing. Clinically, aged cancer patients tend to have a poorer prognosis than young. This may be due to accumulated cellular damage, decreases in adaptive immunity, and chronic inflammation. However, the effects of the aged microenvironment on tumour progression have been largely unexplored. Since dermal fibroblasts can have profound impacts on melanoma progression, we examined whether age-related changes in dermal fibroblasts could drive melanoma metastasis and response to targeted therapy. Here we find that aged fibroblasts secrete a Wnt antagonist, sFRP2, which activates a multi-step signalling cascade in melanoma cells that results in a decrease in β-catenin and microphthalmia-associated transcription factor (MITF), and ultimately the loss of a key redox effector, APE1. Loss of APE1 attenuates the response of melanoma cells to DNA damage induced by reactive oxygen species, rendering the cells more resistant to targeted therapy (vemurafenib). Age-related increases in sFRP2 also augment both angiogenesis and metastasis of melanoma cells. These data provide an integrated view of how fibroblasts in the aged microenvironment contribute to tumour progression, offering new possibilities for the design of therapy for the elderly.
Sigma-Aldrich
Anti-SFRP2 Antibody, clone 80.8.6, azide free, clone 80.8.6, from mouse