MilliporeSigma
  • Indium-bipyridine-catalyzed, enantioselective thiolysis of meso-epoxides.
Indium-bipyridine-catalyzed, enantioselective thiolysis of meso-epoxides.
Chemical communications (Cambridge, England) (2007-06-28)
Mecheril Valsan Nandakumar, Andreas Tschöp, Harald Krautscheid, Christoph Schneider
ABSTRACT
The indium-bipyridine-catalyzed, enantioselective ring-opening of meso-epoxides with aliphatic and aromatic thiols furnished 1,2-mercapto alcohols in good yields and excellent enantioselectivities; the crystal structure of the chiral catalyst reveals a pentagonal-bipyramidal coordination geometry around the indium center.
Sigma-Aldrich
Indium(III) chloride, 99.999% trace metals basis