Merck
  • High efficiency dye-sensitized nanocrystalline solar cells based on ionic liquid polymer gel electrolyte.
High efficiency dye-sensitized nanocrystalline solar cells based on ionic liquid polymer gel electrolyte.
Chemical communications (Cambridge, England) (2003-01-23)
Peng Wang, Shaik M Zakeeruddin, Ivan Exnar, Michael Grätzel
ABSTRACT
An ionic liquid polymer gel containing 1-methyl-3-propylimidazolium iodide (MPII) and poly(vinylidenefluoride-co-hexafluoropropylene) (PVDF-HFP) has been employed as quasi-solid-state electrolyte in dye-sensitized nanocrystalline TiO2 solar cells with an overall conversion efficiency of 5.3% at AM 1.5 illumination.
Sigma-Aldrich
1-Methyl-3-propylimidazolium iodide, ≥98.0% (HPLC)