MilliporeSigma
  • Cytoskeletal regulation by AUTS2 in neuronal migration and neuritogenesis.
Cytoskeletal regulation by AUTS2 in neuronal migration and neuritogenesis.
Cell reports (2014-12-24)
Kei Hori, Taku Nagai, Wei Shan, Asami Sakamoto, Shinichiro Taya, Ryoya Hashimoto, Takashi Hayashi, Manabu Abe, Maya Yamazaki, Keiko Nakao, Tomoki Nishioka, Kenji Sakimura, Kiyofumi Yamada, Kozo Kaibuchi, Mikio Hoshino
ABSTRACT
Mutations in the Autism susceptibility candidate 2 gene (AUTS2), whose protein is believed to act in neuronal cell nuclei, have been associated with multiple psychiatric illnesses, including autism spectrum disorders, intellectual disability, and schizophrenia. Here we show that cytoplasmic AUTS2 is involved in the regulation of the cytoskeleton and neural development. Immunohistochemistry and fractionation studies show that AUTS2 localizes not only in nuclei, but also in the cytoplasm, including in the growth cones in the developing brain. AUTS2 activates Rac1 to induce lamellipodia but downregulates Cdc42 to suppress filopodia. Our loss-of-function and rescue experiments show that a cytoplasmic AUTS2-Rac1 pathway is involved in cortical neuronal migration and neuritogenesis in the developing brain. These findings suggest that cytoplasmic AUTS2 acts as a regulator of Rho family GTPases to contribute to brain development and give insight into the pathology of human psychiatric disorders with AUTS2 mutations.
Sigma-Aldrich
Anti-GABA antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-AUTS2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution