MilliporeSigma
  • Synthesis and biological evaluation of naphthoquinone analogs as a novel class of proteasome inhibitors.
Synthesis and biological evaluation of naphthoquinone analogs as a novel class of proteasome inhibitors.
Bioorganic & medicinal chemistry (2010-07-14)
Harshani R Lawrence, Aslamuzzaman Kazi, Yunting Luo, Robert Kendig, Yiyu Ge, Sanjula Jain, Kenyon Daniel, Daniel Santiago, Wayne C Guida, Saïd M Sebti
ABSTRACT
Screening of the NCI Diversity Set-1 identified PI-083 (NSC-45382) a proteasome inhibitor selective for cancer over normal cells. Focused libraries of novel compounds based on PI-083 chloronaphthoquinone and sulfonamide moieties were synthesized to gain a better understanding of the structure-activity relationship responsible for chymotrypsin-like proteasome inhibitory activity. This led to the demonstration that the chloronaphthoquinone and the sulfonamide moieties are critical for inhibitory activity. The pyridyl group in PI-083 can be replaced with other heterocyclic groups without significant loss of activity. Molecular modeling studies were also performed to explore the detailed interactions of PI-083 and its derivatives with the beta5 and beta6 subunits of the 20S proteasome. The refined model showed an H-bond interaction between the Asp-114 and the sulfonamide moiety of the PI-083 in the beta6 subunit.