Merck
  • Hepatic disposition of ximelagatran and its metabolites in pig; prediction of the impact of membrane transporters through a simple disposition model.
Hepatic disposition of ximelagatran and its metabolites in pig; prediction of the impact of membrane transporters through a simple disposition model.
Pharmaceutical research (2010-02-09)
Erik Sjögren, Ulf Bredberg, Erik Allard, Björn Arvidsson, Jonas Bergquist, Tommy B Andersson, Hans Lennernäs
ABSTRACT
The double prodrug, ximelagatran, is bioconverted, via the intermediates ethylmelagatran and N-hydroxymelagatran, to the direct thrombin inhibitor, melagatran. The primary aim of this study was to investigate the hepatic metabolism and disposition of ximelagatran and the intermediates in pig. A secondary aim was to explore a simple in vitro methodology for quantitative investigations of the impact of membrane transporters on the disposition of metabolized drugs. Porcine S1 (supernatant fraction obtained by centrifuging at 1,000 g for 10 min) liver fractions and hepatocytes were incubated in the absence and presence of known membrane transporter inhibitors. The in vitro kinetics and disposition were determined by simultaneously fitting the disappearance of ximelagatran and the appearance of the metabolites. In S1 liver fractions, the metabolism was significantly inhibited by co-incubation of verapamil or ketoconazole, but not by erythromycin, quinine or quinidine. The disposition of ximelagatran and the intermediate metabolites in hepatocytes were influenced, to various degrees, by carrier-mediated transport processes. This work demonstrates that it is possible to obtain profound information on the general mechanisms that are important in the drug liver disposition using the combination of common in vitro systems and the simple disposition model proposed in this study.
Sigma-Aldrich
Ximelagatran, ≥98% (HPLC)