Merck
  • Carbon nanotubes functionalized by click chemistry as scaffolds for the preparation of electrochemical immunosensors. Application to the determination of TGF-beta 1 cytokine.
Carbon nanotubes functionalized by click chemistry as scaffolds for the preparation of electrochemical immunosensors. Application to the determination of TGF-beta 1 cytokine.
The Analyst (2016-07-08)
E Sánchez-Tirado, A González-Cortés, P Yáñez-Sedeño, J M Pingarrón
ABSTRACT
An electrochemical immunosensor for the determination of the multifunctional Transforming Growth Factor β1 (TGF-β1) cytokine has been prepared using multi-walled carbon nanotube (MWCNT)-modified screen-printed carbon electrodes. MWCNTs were functionalized by means of copper(i) catalyzed azide-alkyne cycloaddition ("click" chemistry) as an efficient strategy for the covalent immobilization of immunoreagents without altering their configurations and preserving their biological activity. Alkyne-functionalized IgGs were also prepared and used to assemble IgG-alkyne-azide-MWCNT conjugates used as scaffolds for the immunosensor preparation. After a blocking step with casein, anti-TGF was immobilized and the target cytokine was sandwiched with biotinylated anti-TGF labeled with poly-HRP-labeled streptavidin. The affinity reaction was monitored amperometrically at -0.20 V using the hydroquinone (HQ)/H
Sigma-Aldrich
Sera human, lyophilized powder (from clotted whole blood)