MilliporeSigma
  • Neurotrophin receptors and selective loss of cholinergic neurons in Alzheimer disease.
Neurotrophin receptors and selective loss of cholinergic neurons in Alzheimer disease.
Molecular and chemical neuropathology (1996-05-01)
F Boissière, S Lehéricy, O Strada, Y Agid, E C Hirsch
ABSTRACT
The most consistent neuropathological finding in Alzheimer disease (AD) is the loss of cholinergic neurons of the nucleus basalis of Meynert (NbM). Using immunohistochemistry, we have previously shown that cholinergic neurons located in the ventral striatum were affected, whereas those of the caudate nucleus, putamen, and mesencephalon were spared. Since cholinergic neurons that degenerate in AD are sensitive to NGF and those that are spared are not, it has been hypothesized that the loss of neurotrophins receptors may play a role in the death of cholinergic neurons in AD. Using immunohistochemistry, we have detected the presence of TrkA on most cholinergic neurons from the NbM, on some from those of the striatum, but not on those of the mesencephalon in the human brain. In AD patients, the number of neurons that expressed TrkA was markedly decreased in the NbM very likely as a consequence of cholinergic neuronal loss. In the striatum, despite the loss of high-affinity NGF binding previously reported, no loss of TrkA was observed. Taken together, these results suggest a decreased expression of NGF receptors on the striatal cholinergic neurons in AD. This loss may contribute, when it reaches a crucial threshold, to the death of cholinergic neurons occurring in AD.
Sigma-Aldrich
TRKA (440-end), active, GST tagged human, PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution