Merck
  • Spherical gold nanoparticles and gold nanorods for the determination of gentamicin.
Spherical gold nanoparticles and gold nanorods for the determination of gentamicin.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2016-05-01)
Jarol R Miranda-Andrades, Aurora Pérez-Gramatges, Omar Pandoli, Eric C Romani, Ricardo Q Aucélio, Andrea R da Silva
ABSTRACT
Gentamicin is an antibiotic indicated to treat mastitis in dairy cattle and for the treatment of bacterial resistance in the context of hospital infections. The effect caused by gentamicin on the optical properties of gold nanoparticles aqueous dispersions were used to develop quantitative methods to determine this antibiotic. Two different aqueous dispersions, one containing spherical Au nanoparticles (AuNPs) and the other containing Au nanorods (AuNRs), had their conditions adjusted to enable a stable and sensitive response towards gentamicin. The use of AuNPs, with measurement at 681nm of the rising coupling plasmon band, enabled a limit of detection (LOD) of 0.4ngmL
Sigma-Aldrich
Sodium citrate monobasic, BioXtra, anhydrous, ≥99.5% (T)