MilliporeSigma
Structure-conserving spontaneous transformations between nanoparticles.
Nature communications (2016-11-11)
K R Krishnadas, Ananya Baksi, Atanu Ghosh, Ganapati Natarajan, Thalappil Pradeep
ABSTRACT
Ambient, structure- and topology-preserving chemical reactions between two archetypal nanoparticles, Ag
Sigma-Aldrich
2,4-Dimethylbenzenethiol, 95%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
In Stock
Details...
$144.00