MilliporeSigma
  • Establishment of an immortalized human endometrial stromal cell line with functional responses to ovarian stimuli.
Establishment of an immortalized human endometrial stromal cell line with functional responses to ovarian stimuli.
Reproductive biology and endocrinology : RB&E (2011-08-02)
Munehiro Yuhki, Takashi Kajitani, Takakazu Mizuno, Yuko Aoki, Tetsuo Maruyama
ABSTRACT
Studies on the mechanisms of decidualization and endometriosis are often hampered by lack of primary endometrial cells. To facilitate in vitro studies, we established a human endometrial stromal cell line, KC02-44D, immortalized with human telomerase reverse transcriptase. Upon exposure to ovarian stimuli, KC02-44D cells showed similar cytoskeletal marker or gene expression and biochemical phenotype to primary endometrial stromal cells. KC02-44D would be useful for studies of human endometrial function and its associated pathologies.
Sigma-Aldrich
Monoclonal Anti-Vimentin−Cy3 antibody produced in mouse, clone V9, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
β-Estradiol, ≥98%