MilliporeSigma
  • Immunogenicity of proteome-determined Mycobacterium avium subsp. paratuberculosis-specific proteins in sheep with paratuberculosis.
Immunogenicity of proteome-determined Mycobacterium avium subsp. paratuberculosis-specific proteins in sheep with paratuberculosis.
Clinical and vaccine immunology : CVI (2008-10-11)
Valerie Hughes, John P Bannantine, Susan Denham, Stuart Smith, Alfredo Garcia-Sanchez, Jill Sales, Michael L Paustian, Kevin Mclean, Karen Stevenson
ABSTRACT
Mycobacterium avium subsp. paratuberculosis causes paratuberculosis, a chronic granulomatous enteritis. Detecting animals with paratuberculosis infections is difficult because the currently available tools have low sensitivity and lack specificity; these tools are prone to generating spurious positive test results caused by exposure to environmental M. avium complex organisms. To generate candidate antigens for incorporation into a specific test for paratuberculosis, subspecies-specific proteins were determined by proteomic comparison of M. avium subsp. paratuberculosis and M. avium subsp. avium. Analysis was aimed at revealing proteins only expressed (or predominant) in the protein profile of M. avium subspecies paratuberculosis. Two-dimensional gel electrophoresis resolved approximately 1,000 protein spots from each subspecies. Proteome analysis identified protein spots whose expression profile appeared markedly increased in M. avium subsp. paratuberculosis, and 32 were identified by analysis of their tryptic peptide profile by matrix-assisted laser desorption ionization-time of flight analysis. Thirty of these proteins were cloned, and their recombinant proteins were expressed. Ovine paratuberculosis sera were used to assess their immunoreactivity by enzyme-linked immunosorbent assay (ELISA), Western blotting, and dot blot analysis. Seventeen proteins were detected in at least one of the immunoassays, and eleven proteins were detected by ELISA with an optical density in excess of the cutoff of 0.1 in four of six sera tested. The immunoreactivity of these proteins indicates their potential as unique diagnostic antigens for the development of a specific serological detection of paratuberculosis.
Sigma-Aldrich
Monoclonal Anti-Maltose Binding Protein antibody produced in mouse, clone MBP-17, ascites fluid