MilliporeSigma
  • Biofunctionalized microfiber-assisted formation of intrinsic three-dimensional capillary-like structures.
Biofunctionalized microfiber-assisted formation of intrinsic three-dimensional capillary-like structures.
Tissue engineering. Part A (2014-01-25)
Stefan Weinandy, Simone Laffar, Ronald E Unger, Thomas C Flanagan, Robert Loesel, C James Kirkpatrick, Marc van Zandvoort, Benita Hermanns-Sachweh, Agnieszka Dreier, Doris Klee, Stefan Jockenhoevel
ABSTRACT
A vascular supply network is essential in engineered tissues >100-200-μm thickness. To control vascular network formation in vitro, we hypothesize that capillarization can be achieved locally by using fibers to position and guide vessel-forming endothelial cells within a three-dimensional (3D) matrix. Biofunctionalization of poly-(L-lactic acid) (PLLA) fibers was performed by amino-functionalization and covalent binding of RGD peptides. Human foreskin fibroblasts (HFFs) and human umbilical vein endothelial cells (HUVECs) were seeded on the fibers in a mould and subsequently embedded in fibrin gel. After 9-21 days of coculture, constructs were fixed and immunostained (PECAM-1). Capillary-like structures with lumen in the 3D fibrin matrix were verified and quantified using two-photon microscopy and image analysis software. Capillary-like networks with lumen formed adjacent to the PLLA fibers. Increased cell numbers were observed to attach to RGD-functionalized fibers, resulting in enhanced formation of capillary-like structures. Cocultivation of HFFs sufficiently supported HUVECs in the formation of capillary-like structures, which persisted for at least 21 days of coculture. The guidance of vessel growth within tissue-engineered constructs can be achieved using biofunctionalized PLLA microfibers. Further methods are warranted to perform specified spatial positioning of fibers within 3D formative scaffolds to enhance the applicability of the concept.