MilliporeSigma
  • Lacrimal drainage-associated lymphoid tissue (LDALT): a part of the human mucosal immune system.
Lacrimal drainage-associated lymphoid tissue (LDALT): a part of the human mucosal immune system.
Investigative ophthalmology & visual science (2001-02-27)
E Knop, N Knop
ABSTRACT
Mucosa-associated lymphoid tissue (MALT) specifically protects mucosal surfaces. In a previous study of the human conjunctiva, evidence was also found for the presence of MALT in the lacrimal sac. The present study, therefore, aims to investigate its morphology and topographical distribution in the human lacrimal drainage system. Lacrimal drainage systems (n = 51) obtained from human cadavers were investigated by clearing flat wholemounts or by serial sections of tissue embedded in paraffin, OCT compound, or epoxy resin. These were further analyzed by histology, immunohistochemistry, and electron microscopy. All specimens showed the presence of lymphocytes and plasma cells as a diffuse lymphoid tissue in the lamina propria, together with intraepithelial lymphocytes and occasional high endothelial venules (HEV). It formed a narrow layer along the canaliculi that became thicker in the cavernous parts. The majority of lymphocytes were T cells, whereas B cells were interspersed individually or formed follicular centers. T cells were positive for CD8 and the human mucosa lymphocyte antigen (HML-1). Most plasma cells were positive for IgA and the overlying epithelium expressed its transporter molecule secretory component (SC). Basal mucous glands were present in the lacrimal canaliculi and in the other parts accompanied by alveolar and acinar glands, all producing IgA-rich secretions. Primary and secondary lymphoid follicles possessing HEV were present in about half of the specimens. The term lacrimal drainage-associated lymphoid tissue (LDALT) is proposed here to describe the lymphoid tissue that is regularly present and belongs to the common mucosal immune system and to the secretory immune system. It is suggested that it may form a functional unit together with the lacrimal gland and conjunctiva, connected by tear flow, lymphocyte recirculation, and probably the neural reflex arc, and play a major role in preserving ocular surface integrity.
Sigma-Aldrich
IgA from human colostrum, reagent grade, ≥95%, essentially salt-free, lyophilized powder