MilliporeSigma
  • Disrupted GABAAR trafficking and synaptic inhibition in a mouse model of Huntington's disease.
Disrupted GABAAR trafficking and synaptic inhibition in a mouse model of Huntington's disease.
Neurobiology of disease (2012-03-10)
Eunice Y Yuen, Jing Wei, Ping Zhong, Zhen Yan
ABSTRACT
Growing evidence suggests that Huntington's disease (HD), a neurodegenerative movement disorder caused by the mutant huntingtin (htt) with an expanded polyglutamine (polyQ) repeat, is associated with the altered intracellular trafficking and synaptic function. GABA(A) receptors, the key determinant of the strength of synaptic inhibition, have been found to bind to the huntingtin associated protein 1 (HAP1). HAP1 serves as an adaptor linking GABA(A) receptors to the kinesin family motor protein 5 (KIF5), controlling the transport of GABA(A) receptors along microtubules in dendrites. In this study, we found that GABA(A)R-mediated synaptic transmission is significantly impaired in a transgenic mouse model of HD expressing polyQ-htt, which is accompanied by the diminished surface expression of GABA(A) receptors. Moreover, the GABA(A)R/HAP1/KIF5 complex is disrupted and dissociated from microtubules in the HD mouse model. These results suggest that GABA(A)R trafficking and function is impaired in HD, presumably due to the interference of KIF5-mediated microtubule-based transport of GABA(A) receptors. The diminished inhibitory synaptic efficacy could contribute to the loss of the excitatory/inhibitory balance, leading to increased neuronal excitotoxicity in HD.
Sigma-Aldrich
Monoclonal Anti-Synaptophysin antibody produced in mouse, clone SVP-38, ascites fluid
Sigma-Aldrich
Anti-GABA A Receptor β 2,3 Chain Antibody, clone BD17, clone BD17, Chemicon®, from mouse
Sigma-Aldrich
Anti-Huntingtin Protein Antibody, a.a. 181-810, clone 1HU-4C8, ascites fluid, clone 1HU-4C8, Chemicon®
Sigma-Aldrich
Anti-NR2A Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone DM1A, purified from hybridoma cell culture