Merck
  • Tristetraprolin Down-Regulation Contributes to Persistent TNF-Alpha Expression Induced by Cigarette Smoke Extract through a Post-Transcriptional Mechanism.
Tristetraprolin Down-Regulation Contributes to Persistent TNF-Alpha Expression Induced by Cigarette Smoke Extract through a Post-Transcriptional Mechanism.
PloS one (2016-12-03)
Xue-Ke Zhao, Pulin Che, Ming-Liang Cheng, Quan Zhang, Mao Mu, Hong Li, Yuan Luo, Yue-Dong Liang, Xin-Hua Luo, Chang-Qing Gao, Patricia L Jackson, J Michael Wells, Yong Zhou, Meng Hu, Guoqiang Cai, Victor J Thannickal, Chad Steele, J Edwin Blalock, Xiaosi Han, Ching-Yi Chen, Qiang Ding
ABSTRACT
Tumor necrosis factor-alpha (TNF-α) is a potent pro-inflammatory mediator and its expression is up-regulated in chronic obstructive pulmonary disease (COPD). Tristetraprolin (TTP) is implicated in regulation of TNF-α expression; however, whether TTP is involved in cigarette smoke-induced TNF-α expression has not been determined. TTP expression was examined by western blot analysis in murine alveolar macrophages and alveolar epithelial cells challenged without or with cigarette smoke extract (CSE). TNF-α mRNA stability, and the decay of TNF-α mRNA, were determined by real-time quantitative RT-PCR. TNF-α protein levels were examined at the same time in these cells. To identify the molecular mechanism involved, a construct expressing the human beta-globin reporter mRNA containing the TNF-α 3'-untranslated region was generated to characterize the TTP targeted site within TNF-α mRNA. CSE induced TTP down-regulation in alveolar macrophages and alveolar epithelial cells. Reduced TTP expression resulted in significantly increased TNF-α mRNA stability. Importantly, increased TNF-α mRNA stability due to impaired TTP function resulted in significantly increased TNF-α levels in these cells. Forced TTP expression abrogated the increased TNF-α mRNA stability and expression induced by CSE. By using the globin reporter construct containing TNF-α mRNA 3'-untranslated region, the data indicate that TTP directly targets the adenine- and uridine-rich region (ARE) of TNF-α mRNA and negatively regulates TNF-α expression at the post-transcriptional level. The data demonstrate that cigarette smoke exposure reduces TTP expression and impairs TTP function, resulting in significantly increased TNF-α mRNA stability and excessive TNF-α expression in alveolar macrophages and epithelial cells. The data suggest that TTP is a novel post-transcriptional regulator and limits excessive TNF-α expression and inflammatory response induced by cigarette smoke.
Sigma-Aldrich
Thymidine 5′-triphosphate sodium salt, ≥96%