MilliporeSigma
  • Nitric oxide signaling is involved in the response to iron deficiency in the woody plant Malus xiaojinensis.
Nitric oxide signaling is involved in the response to iron deficiency in the woody plant Malus xiaojinensis.
Plant physiology and biochemistry : PPB (2016-11-12)
Longmei Zhai, Dashuang Xiao, Chaohua Sun, Ting Wu, Zhenhai Han, Xinzhong Zhang, Xuefeng Xu, Yi Wang
ABSTRACT
To cope with iron (Fe) deficiency, plants have evolved a wide range of adaptive responses from changes in morphology to altered physiological responses. Recent studies have demonstrated that nitric oxide (NO) is involved in the Fe-deficiency response through hormonal signaling pathways. Here, we report that NO plays a significant role in Malus xiaojinensis, an Fe-efficient woody plant. Fe deficiency triggered significant accumulation of NO in the root system, predominantly in the outer cortical and epidermal cells of the elongation zone. The NO scavenger 2-(4-carboxyphenyl)-4,4,5,5-tetramethylimidazoline-1-oxyl-3-oxide potassium salt (cPTIO) completely arrested Fe deficiency-induced root hair formation, blocked the increase in root ferric-chelate reductase activity and in root H+ excretion, further reduced the active iron content in young leaves and roots, and prevented the upregulation of the critical Fe-related genes, FIT, MxFRO2-like, and MxIRT1. These conditions were restored under Fe deficiency by treatment with the NO donor, sodium nitroprusside (SNP). Additionally, chlorophyll content and relative expression levels of the genes chlorophyll a deoxygenase (MxCAO) and polyamine oxidase (MxPAO) were not changed significantly following Fe deficiency for 6 d; however, SNP treatment increased MxHEMA gene expression. Interestingly, the Fv/Fm ratio, the maximum quantum yield of photosystem II (PSII), decreased significantly following cPTIO treatment. We observed more severe chlorosis under Fe deficiency with cPTIO treatment for 9 d. These results strongly suggest that NO mediates a range of responses to Fe deficiency in M. xiaojinensis, from morphological changes to the regulation of physiological processes and gene expression.
Sigma-Aldrich
DAF-FM DA, ≥98% (HPLC)