MilliporeSigma
  • Modeling quorum sensing trade-offs between bacterial cell density and system extension from open boundaries.
Modeling quorum sensing trade-offs between bacterial cell density and system extension from open boundaries.
Scientific reports (2016-12-15)
Mattia Marenda, Marina Zanardo, Antonio Trovato, Flavio Seno, Andrea Squartini
ABSTRACT
Bacterial communities undergo collective behavioural switches upon producing and sensing diffusible signal molecules; a mechanism referred to as Quorum Sensing (QS). Exemplarily, biofilm organic matrices are built concertedly by bacteria in several environments. QS scope in bacterial ecology has been debated for over 20 years. Different perspectives counterpose the role of density reporter for populations to that of local environment diffusivity probe for individual cells. Here we devise a model system where tubes of different heights contain matrix-embedded producers and sensors. These tubes allow non-limiting signal diffusion from one open end, thereby showing that population spatial extension away from an open boundary can be a main critical factor in QS. Experimental data, successfully recapitulated by a comprehensive mathematical model, demonstrate how tube height can overtake the role of producer density in triggering sensor activation. The biotic degradation of the signal is found to play a major role and to be species-specific and entirely feedback-independent.
Sigma-Aldrich
N-Octanoyl-L-homoserine lactone, ≥96% (HPLC)
Product 44558 is not currently sold in your country. Contact Technical Service