Merck
Cytosporone B is an agonist for nuclear orphan receptor Nur77.
Nature chemical biology (2008-08-12)
Yanyan Zhan, Xiping Du, Hangzi Chen, Jingjing Liu, Bixing Zhao, Danhong Huang, Guideng Li, Qingyan Xu, Mingqing Zhang, Bart C Weimer, Dong Chen, Zhe Cheng, Lianru Zhang, Qinxi Li, Shaowei Li, Zhonghui Zheng, Siyang Song, Yaojian Huang, Zhiyun Ye, Wenjin Su, Sheng-Cai Lin, Yuemao Shen, Qiao Wu
ABSTRACT
Nuclear orphan receptor Nur77 has important roles in many biological processes. However, a physiological ligand for Nur77 has not been identified. Here, we report that the octaketide cytosporone B (Csn-B) is a naturally occurring agonist for Nur77. Csn-B specifically binds to the ligand-binding domain of Nur77 and stimulates Nur77-dependent transactivational activity towards target genes including Nr4a1 (Nur77) itself, which contains multiple consensus response elements allowing positive autoregulation in a Csn-B-dependent manner. Csn-B also elevates blood glucose levels in fasting C57 mice, an effect that is accompanied by induction of multiple genes involved in gluconeogenesis. These biological effects were not observed in Nur77-null (Nr4a1-/-) mice, which indicates that Csn-B regulates gluconeogenesis through Nur77. Moreover, Csn-B induced apoptosis and retarded xenograft tumor growth by inducing Nur77 expression, translocating Nur77 to mitochondria to cause cytochrome c release. Thus, Csn-B may represent a promising therapeutic drug for cancers and hypoglycemia, and it may also be useful as a reagent to increase understanding of Nur77 biological function.
Sigma-Aldrich
9-cis-Retinoic acid, ≥98% (HPLC)