MilliporeSigma
  • Hepatitis C virus has a genetically determined lymphotropism through co-receptor B7.2.
Hepatitis C virus has a genetically determined lymphotropism through co-receptor B7.2.
Nature communications (2017-01-10)
Chia-Lin Chen, Jeffrey Y Huang, Chun-Hsiang Wang, Stanley M Tahara, Lin Zhou, Yasuteru Kondo, Joel Schechter, Lishan Su, Michael M C Lai, Takaji Wakita, François-Loïc Cosset, Jae U Jung, Keigo Machida
ABSTRACT
B-cell infection by hepatitis C virus (HCV) has been a controversial topic. To examine whether HCV has a genetically determined lymphotropism through a co-receptor specific for the infection by lymphotropic HCV, we established an infectious clone and chimeric virus of hepatotropic and lymphotropic HCV strains derived from an HCV-positive B-cell lymphoma. The viral envelope and 5'-UTR sequences of the lymphotropic HCV strain were responsible for the lymphotropism. Silencing of the virus sensor, RIGI, or overexpression of microRNA-122 promoted persistent viral replication in B cells. By cDNA library screening, we identified an immune cell-specific, co-stimulatory receptor B7.2 (CD86) as a co-receptor of lymphotropic HCV. Infection of B cells by HCV inhibited the recall reaction to antigen stimulation. Together, a co-receptor B7.2 enabled lymphotropic HCV to infect memory B cells, leading to inhibition of memory B-cell function and persistent HCV infection in HCV-infected hosts.
Sigma-Aldrich
MISSION® esiRNA, targeting human IFIH1
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
0.25 mg
In Stock
Details...
$225.00