MilliporeSigma
  • A two-step synthetic strategy to obtain a water-soluble derivative of curcumin with improved antioxidant capacity and in vitro cytotoxicity in C6 glioma cells.
A two-step synthetic strategy to obtain a water-soluble derivative of curcumin with improved antioxidant capacity and in vitro cytotoxicity in C6 glioma cells.
Materials science & engineering. C, Materials for biological applications (2016-12-19)
José M Landeros, Fernando Belmont-Bernal, Alma Teresa Pérez-González, Mario Israel Pérez-Padrón, Patricia Guevara-Salazar, Irma Gabriela González-Herrera, Patricia Guadarrama
ABSTRACT
A novel water-soluble derivative of curcumin (Cur-[G-2]-OH) was designed and synthesized from accessible raw materials in only two steps with an overall yield of 80%. The modification of curcumin phenol groups with second-generation polyester dendrons (dendronization) as a strategy to achieve an optimal hydrophilic/hydrophobic balance allows the complete water solubilization of the new curcumin derivative (5mg/ml) at room temperature. The therapeutic potential of Cur-[G-2]-OH was investigated in terms of antioxidant capacity, intracellular uptake and cytotoxicity in both rat glioblastoma cells and normal human dermal fibroblasts. Although the phenolic groups of curcumin were locked by dendronization, Cur-[G-2]-OH exhibited antioxidant capacity in water that was even higher than curcumin in dimethylsulfoxide (DMSO). This compound showed a steady cellular uptake contrasted with curcumin, which has a saturation capture at high concentrations. Combined with improved stability, this property seems to allow the intracellular accumulation of Cur-[G-2]-OH. Furthermore, the new compound exhibited increased cytotoxicity in rat C6 glioma cells in a time- and concentration-dependent manner, whereas in normal human fibroblasts, its IC
Sigma-Aldrich
2,2-Bis(hydroxymethyl)propionic acid, 98%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 mL
In Stock
Details...
$213.00
1 L
Estimated to ship on March 24, 2025
$274.00
2 L
Estimated to ship on March 24, 2025
$343.00
Sigma-Aldrich
Benzyl bromide, reagent grade, 98%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 mL
In Stock
Details...
$213.00
1 L
Estimated to ship on March 24, 2025
$274.00
2 L
Estimated to ship on March 24, 2025
$343.00