MilliporeSigma
  • New investigational drugs with single-agent activity in multiple myeloma.
New investigational drugs with single-agent activity in multiple myeloma.
Blood cancer journal (2016-07-30)
A M Rajan, S Kumar
ABSTRACT
The treatment of multiple myeloma (MM) is rapidly evolving. In the United States, four drugs (panobinostat, ixazomib, daratumumab and elotuzumab) were approved for the treatment of MM in 2015. As a result of improved diagnosis and therapy, there has been a dramatic improvement in the outcome of MM in the last decade, probably more than any other malignancy. Numerous agents continue to be studied in preclinical models and in clinical trials, with many demonstrating clinical efficacy that appears promising enough to have a trajectory for regulatory approval. The purpose of this article is to summarize the current data and provide perspective on new investigational agents with promising single-agent activity in MM. The agents reviewed include Isatuximab, an anti-CD38 monoclonal antibody; marizomib, a new proteasome inhibitor; oprozomib, an oral proteasome inhibitor; filanesib (ARRY-520), a kinesin spindle protein inhibitor; dinaciclib, a cyclin-dependent kinase inhibitor; venetoclax (ABT-199), a selective BCL-2 inhibitor; and LGH-447, pan PIM kinase inhibitor.
Sigma-Aldrich
Marizomib, ≥95% (HPLC), (Salinospora tropica)