MilliporeSigma
  • Novel biocomposite of carboxymethyl chitosan and pineapple peel carboxymethylcellulose as sunscreen carrier.
Novel biocomposite of carboxymethyl chitosan and pineapple peel carboxymethylcellulose as sunscreen carrier.
International journal of biological macromolecules (2016-10-26)
Lucksanee Wongkom, Ampa Jimtaisong
ABSTRACT
This study aims to prepare of biocomposite of carboxymethyl chitosan (CM-chitosan) and carboxymethylcellulose (CMC) from Ananas comosus (pineapple) peel for use as broad spectrum sunscreen carrier. Biocomposite was produced by using ferulic acid (FA), a plant extract, as crosslinker with the optimal ratio of CMC: CM-chitosan: FA at 1:2:4%w. FT-IR technique demonstrated that crosslinking may occur at amine group of CM-chitosan and carboxyl group of FA and hydrogen bonding between hydroxyl group of CMC and carboxyl group of FA. Biocomposite is pale yellow powder and present fibre bundle-like surface in the SEM image. DSC, TGA and XRD results indicated that new compound was formed. The particle size of biocomposite is 626nm determined by using Zetasizer. Hydrophilic TiO
Sigma-Aldrich
2-Phenylbenzimidazole, 97%